Få mere at vide
Helidaminic Acid, TRC
Artikelnummer. 30395721
Skift visning
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30395721 10 g
30408579 1 g
30406921 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30395721 Leverandør Toronto Research Chemicals Leverandørnr. TRCH23513310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 499-51-4
Molekylær formel C7H5NO5
Molekylvægt (g/mol) 183.12
Synonym 4-Hydroxy-2,6-pyridinedicarboxylic Acid, 2,6-Dicarboxy-4-hydroxypyridine, 4-Hydroxy-2,6-dicarboxypyridine, 4-Hydroxydipicolinic Acid, 4-Hydroxypyridine-2,6-carboxylic Acid, 4-Hydroxypyridine-2,6-dicarboxylic Acid
SMIL OC(C1=CC(O)=CC(C(O)=O)=N1)=O
Tekniske data
Kemisk navn eller materiale Helidaminic Acid
Mængde 10 g
InChI formel InChI=1S/C7H5NO5/c9-3-1-4(6(10)11)8-5(2-3)7(12)13/h1-2H,(H,8,9)(H,10,11)(H,12,13)