missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ HEC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578318
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Positive Control: HeLa, Blocking peptide (GTX70014-PEP) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Tekniske data
| HEC1 | |
| Polyclonal | |
| Unconjugated | |
| NDC80 | |
| 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10403 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.816 mg/mL | |
| PBS with no preservative | |
| O14777 | |
| NDC80 | |
| Synthetic peptide corresponding to the amino acids around Ser 76/77 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion