Få mere at vide
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-44
Artikelnummer. 17746973 Shop alle 3M produkter
:
Breathing Tube; For Use With: 3M™ PAPR Systems
Artikelnummer. 17746973
Brand: 3M™ 7100236824
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-44 is a fixed-length EPDM rubber breathing tube designed for use with certain approved 3M™ Powered Air Systems.
  • Fixed-length breathing tube made from EPDM rubber
  • Approved for use with 3M™ Powered Air Respirator PF-600E
  • Suitable for use with specific 3M™ Versaflo™ headtops when using the 3M™ Adapter Assembly QRS Thread ADP-03.
  • 80 cm length
TRUSTED_SUSTAINABILITY
Tekniske data
Materiale EPDM Rubber