Få mere at vide
3M™ Heavy Duty EPDM Breathing Tube
Artikelnummer. 17796993 Shop alle 3M produkter
Model-:
SC-BT-44-L
Artikelnummer. 17796993
Brand: 3M™ 7100265244
Extended fixed-length breathing tube made from EPDM rubber.
  • Suitable for use with specific 3M™ Versaflo™ headtops including M-200, M-300 and S Series Hoods.
  • EPDM rubber hose
  • 100 cm length
  • Protective breathing tube covers available to lengthen service life of breathing tubes