Beskrivelse
- Cut time and waste and ensure staff have as many cleaning cloths as they need close by, thanks to the portable Handy Box.
- Optimise consumption of our disposable cleaning cloths with the one-at-a-time dispensing feature.
- The Handy Box's large opening means cloths are easy to access, even when wearing gloves.
- Improve your brand's sustainability - our packaging is made out of 100% recycled fibres.
- Folded length between 10.5 cm to 10.8 cm.
Tekniske data
Tekniske data
| Produkttype | Heavy-duty Wiper |
| Bredde (engelsk) | 13.98 in. |
| Bredde (metrisk) | 35.5 cm |
| Farve | Hvid |
| Emballagemængde | 120 stk./pakke |
| Ply | 1-ply |