missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEATR4 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05832JF549
Additional Details : Weight : 0.00970kg
Description
HEATR4 Polyclonal specifically detects HEATR4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HEATR4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
HEAT repeat containing 4, HEAT repeat-containing protein 4, MGC48595 | |
Produced in rabbits immunized with E. coli-derived Human HEATR4 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
399671 | |
Store at 4°C in the dark. | |
IgG |