Learn More
Grant Instruments™ Heating Block for TS-DW Deep-well Microplate Thermoshaker
Accommodates one deep-well plate for TS-DW Deep-well Microplate Thermoshaker
Brand: Grant Instruments™ B-2A
5005.57 DKK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
Block for one deep-well plate- Shaped to fit the microplates for better heat transfer
- Three year warranty
Specifications
TS-DW Microplate Thermoshaker | |
Block | |
Axygen™ 96/2200µl deep-well plate |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%