Learn More
Kautex™ HDPE Natural Rectangular Narrow Neck Bottle
Brand: Kautex™ 2000070520
Additional Details : Weight : 0.97200kg
Description
- High stability, ribbed sides for secure grip
- Rectangular, natural color
- Caps are not included; sold separately.
Specifications
Narrow Neck Bottle | |
0.1 L | |
12.9 mm | |
HDPE | |
No | |
Square |
110.5 mm | |
Natural | |
Storage, Packaging, Dispensing, Mailing | |
Narrow | |
Narrow |