Learn More
VITLAB™ HDPE Measuring Scoop
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ HDPE Measuring Scoops are lighter than metal, don’t dent and feature good chemical resistance.
Brand: VITLAB™ 40293
This item is not returnable.
View return policy
Description
- Conical in shape with tapered filling edge
Specifications
Measuring Scoop | |
Conical | |
Natural |
HDPE | |
65 mL | |
Suitable as weighing |
Product Content Correction
Product Title
VITLAB™ HDPE Measuring Scoop > Capacity: 65mL
Spot an opportunity for improvement?Share a Content Correction