Learn More
Buerkle™ HDPE, Compact Jerry Can (Electroconductive) with Threaded Connector
For Use With: For use in explosion-proof areas; Closure Material: HDPE; Color: Black
528.00 DKK - 692.00 DKK
Specifications
Closure Color | Black |
---|---|
Closure Material | HDPE |
Color | Black |
Description | Canister |
Disposable | Reusable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
16319541
|
Buerkle™
0435-2055 |
5 L |
528.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
16329541
|
Buerkle™
0435-2105 |
10 L |
692.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
Description
- HDPE, black, anti-static
- For use in explosion-proof areas
- For dispensing and storing combustible and highly flammable liquids
- With or without threaded connector 3/4 in.
- Due to its narrow width, the canister requires very little room
Specifications
Black | |
Black | |
Reusable | |
StopCock PA | |
13.19 in. | |
13.19 in. | |
Anti-static HDPE | |
Jerry Can |
HDPE | |
Canister | |
For Use in Explosion-Proof Areas | |
Liter | |
335 mm | |
335 mm | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Buerkle™ HDPE, Compact Jerry Can (Electroconductive) with Threaded Connector