missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS (10X), calcium, magnesium, phenol red

HBSS (10X), calcium, magnesium, phenol red

288.00 DKK - 2505.00 DKK

Specifications

Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 15 to 30°C
Concentration 10 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11510466
View Documents Promotion Details
Gibco™
14060040
500 mL
288.00 DKK
500mL
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11520466
View Documents Promotion Details
Gibco™
14060073
10 x 500 mL
2505.00 DKK
5000mL
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Concentrated forms of Gibco™ HBSS require pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications

Specifications

ISO 13485 standards, FDA
10 X
Hanks' Balanced Salt Solution
No Sodium Bicarbonate
5.5 to 6.0
15 to 30°C
Liquid
Calcium, Magnesium, Glucose, Phenol Red
Mammalian Cell Culture
For in-vitro diagnostic use
Videos
SDS
Documents

Documents

Product Certifications