missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS (10X), calcium, magnesium, phenol red

HBSS (10X), calcium, magnesium, phenol red

Brand:  Gibco™ 14060073

Additional Details : Weight : 0.52500kg

 View more versions of this product

Product Code. 11520466

  • 2035.00 DKK / 5000mL

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Concentrated forms of Gibco™ HBSS require pH adjustment and filtration at the time of preparation (see protocol for details).
Specifications

Specifications

10 x
With - Glucose, sodium bicarbonate, phenol red, without - calcium, magnesium
For in-vitro diagnostic use
15 to 30°C
10 x 500mL
ISO 13485 standards, FDA
Calcium, magnesium, glucose, phenol red
Sodium bicarbonate
Hanks' Balanced Salt Solution, 10 x 500mL, Liquid, 10X Concentration, 5.5 to 6.0 pH, With Calcium, Magnesium, Glucose, Phenol Red, 36 Months Shelf Life, For Mammalian Cell Culture
Hanks' Balanced Salt Solution
15 to 30°C
36 Months
Liquid
Liquid
5.5 to 6.0
Mammalian cell culture
Videos
SDS
Documents