Learn More
Invitrogen™ Human HAX1 alpha Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0769
Additional Details : Weight : 0.01000kg
Description
Hax1a Synthetic Peptide, PEP-0769, from Invitrogen.
This peptide corresponds to 15 amino acids near the amino terminus of human Hax1 isoform a.PEP-0769 can be used as a blocking peptide with polyclonal antibody PA5-20649.Specifications
10456 | |
purified | |
Human HAX1 alpha | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O00165 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
HAX1 | |
0.02% sodium azide | |
50 μg | |
PBS |