missing translation for 'onlineSavingsMsg'
Få mere at vide

Harpagide (90%), TRC

Artikelnummer. 30399479
Change view
Click to view available options
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30399479 100 mg
30390993 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399479 Supplier Toronto Research Chemicals Supplier No. TRCH105270100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6926-08-5
Molekylær formel C15 H24 O10
Molekylvægt (g/mol) 364.35
Synonym β-D-Glucopyranoside, (1S,4aS,5R,7S,7aR)-1,4a,5,6,7,7a-hexahydro-4a,5,7-trihydroxy-7-methylcyclopenta[c]pyran-1-yl, Glucopyranoside, 1β,4a,5,6,7,7aα-hexahydro-4aα,5α,7α-trihydroxy-7-methylcyclopenta[c]pyran-1-yl, β-D- (8CI), Harpagide (7CI), β-D-Glucopyranoside, 1,4a,5,6,7,7a-hexahydro-4a,5,7-trihydroxy-7-methylcyclopenta[c]pyran-1-yl, [1S-(1α,4aα,5α,7α,7aα)]-, (1S,4aS,5R,7S,7aR)-1,4a,5,6,7,7a-Hexahydro-4a,5,7-trihydroxy-7-methylcyclopenta[c]pyran-1-yl β-D-glucopyranoside
IUPAC navn (1S,4aS,5R,7S,7aR)-7-methyl-1-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1,5,6,7a-tetrahydrocyclopenta[c]pyran-4a,5,7-triol
SMIL C[C@]1(O)C[C@@H](O)[C@]2(O)C=CO[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H]12

Tekniske data

Kemisk navn eller materiale Harpagide
Mængde 100 mg
InChI formel InChI=1S/C15H24O10/c1-14(21)4-7(17)15(22)2-3-23-13(11(14)15)25-12-10(20)9(19)8(18)6(5-16)24-12/h2-3,6-13,16-22H,4-5H2,1H3/t6-,7-,8-,9+,10-,11-,12+,13+,14+,15-/m1/s1
Formel vægt 364.1369
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.