Få mere at vide
Harmine-d3, TRC
Artikelnummer. 30404460
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30404460 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404460 Leverandør Toronto Research Chemicals Leverandørnr. TRCH10525210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Harmine-d3
CAS 1216704-96-9
Molekylær formel C13H9D3N2O
Mængde 10 mg
InChI formel InChI=1S/C13H12N2O/c1-8-13-11(5-6-14-8)10-4-3-9(16-2)7-12(10)15-13/h3-7,15H,1-2H3/i2D3
SMIL [2H]C([2H])([2H])Oc1ccc2c(c1)[nH]c3c(C)nccc23
Molekylvægt (g/mol) 215.27
Formel vægt 215.1138