Learn More
RETSCH Hardened Stainless Steel O-ring
Brand:  RETSCH 05.114.0054
2113.75 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.00400kg
Product Code. 10486132
  • 2225.00 DKK / Each
Estimated Shipment: 26-08-2024
to see stock.
Specifications
O-Ring
1Each
Hardened Stainless Steel
For Planetary Ball Mill PM 100 CM 500mL Grinding Jars