Få mere at vide
3M™ Hard Hat Series G2001 Dielectric 1000v
Shop alle 3M produkter
:
Color: White
Beskrivelse
- A safety helmet with excellent ventilation a good field of vision and one of few lightweight helmets available
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it
- Extended edge on the back of the helmet provides extra protection for the sensitive neck area
- Flat surface for printing a logotype, brand name, etc
- Easy to replace sweatband for comfort and hygiene
- Electrical insulation to 1000Vac
Tekniske data
Tekniske data
| Certificeringer/compliance | EN 50365 |
| Farve | White |
| Inkluderer | Leather Sweatband |
| Materiale | UV-stabilized ABS |
| Produkttype | Helmet |
| Størrelse | 54 to 62 cm |
| Vægt (metrisk) | 340 g |
| Ventileret | Not Ventilated |