Learn More
Hamilton™ 700 Microliter™ Syringes: RN Termination
Product Code. 10363851
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Gauge:
22 G
22s G
26s G
Model:
75
701
702
705
710
725
750
Quantity:
6/Pk.
Ea.
Unit Size:
Each
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Model Quantity Unit Size
10363851 10 μL 26s G 701 Ea. Each
10078460 25 μL 22s G 702 Ea. Each
10130411 50 μL 22s G 705 Ea. Each
10292151 100 μL 22s G 710 Ea. Each
10528321 250 μL 22s G 725 Ea. Each
10528511 500 μL 22 G 750 Ea. Each
10136301 10 μL 26s G 701 6/Pk. Pack of 6
10078450 5 μL 26s G 75 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10363851 Supplier Hamilton™ Supplier No. 80330
  • ONLINE EXCLUSIVE PRICE
    752.00 DKK
    667.98 DKK / Pakke med 1
    Spare 84.02 DKK
    11% Slukket
Forventet forsendelse: 29-12-2026
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Material PTFE (Tip), Stainless Steel (Plunger)
Needle Gauge 26s G
Diameter (English) Inner 0.019 in.
Diameter (English) Inner Needle 0.005 in.
Diameter (Metric) Inner Needle 0.13 mm
Diameter (English) Outer Needle 0.0187 in.
Diameter (English) Outer 0.26 in.
Diameter (Metric) Inner 0.485 mm
Diameter (Metric) Outer 6.6 mm
Diameter (Metric) Outer Needle 0.47 mm
For Use With (Application) For dispensing low volumes to achieve the highest level of accuracy and precision possible
Length (English) 2 in.
Length (English) Needle 2 in.
Length (Metric) 51 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 239°F
Max. Temperature (Metric) 115°C
Min. Temperature (English) 50°F
Model 701
Needle Point Number 2
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Removable