Få mere at vide
Halobetasol, TRC
Artikelnummer. 30137412
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137412 100 mg
30003563 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137412 Leverandør Toronto Research Chemicals Leverandørnr. TRCH102290100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 98651-66-2
Molekylær formel C22 H27 Cl F2 O4
Molekylvægt (g/mol) 428.9
Synonym Pregna-1,4-diene-3,20-dione, 21-chloro-6,9-difluoro-11,17-dihydroxy-16-methyl-, (6α,11β,16β)-, (6α,11β,16β)-21-Chloro-6,9-difluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, Halobetasol, Ulobetasol
IUPAC navn (6S,8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-6,9-difluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMIL C[C@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CCl
Tekniske data
Kemisk navn eller materiale Ulobetasol
Mængde 100 mg
InChI formel InChI=1S/C22H27ClF2O4/c1-11-6-13-14-8-16(24)15-7-12(26)4-5-19(15,2)21(14,25)17(27)9-20(13,3)22(11,29)18(28)10-23/h4-5,7,11,13-14,16-17,27,29H,6,8-10H2,1-3H3/t11-,13-,14-,16-,17-,19-,20-,21-,22-/m0/s1
Formel vægt 428.1566