Få mere at vide
Invitrogen™ HAL Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542754
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Peptide sequence: INKLQELQVN LVRSHSSGVG KPLSPERCRM LLALRINVLA KGYSGISLET Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.
Tekniske data
| HAL | |
| Polyclonal | |
| Unconjugated | |
| HAL | |
| hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3034 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P42357 | |
| HAL | |
| Synthetic peptide directed towards the N-terminal of human HAL (aa 180-229). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |