missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Hairy (Drosophila) Antibody (1/24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76659
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Tekniske data
| Hairy (Drosophila) | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot : 1:100 - 1:2000, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Mouse | |
| Protein A purified | |
| Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells | |
| This antibody will stain Drosophila embryos. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, ChIP Assay | |
| 1/24 | |
| Unconjugated | |
| PBS with 0.02% sodium azide | |
| Hairy gene product | |
| 0.1 mL | |
| Primary | |
| Drosophila | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion