missing translation for 'onlineSavingsMsg'
Learn More

Hafnium solution for ICP 10g/L HCl/HF matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Brand:  Fisher Chemical S223250

Additional Details : Weight : 0.25000kg

Packaging Bottle
Quantity 250mL

 View more versions of this product

Product Code. 12906314

  • 6957.00 DKK / 250mL

Estimated Shipment
19-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

Specifications

10±0.02g/L
Hf
250mL
Bottle
HCl/HF
Mass spectrometry standards
Videos
SDS
Documents