Få mere at vide
MedChemExpress HAEGT
Artikelnummer. 16412859
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16412859 1 mg
1 options
Artikelnummer. 16412859 Leverandør MedChemExpress Leverandørnr. HYP1230
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
MedChemExpress HAEGT is the first N-terminal 1-5 residues of glucagon like peptide-1 (GLP-1) peptide, and the sequence is His-Ala-Glu-Gly-Thr. HAEGT acts as a competitive substrate for probing prime substrate binding sites of human dipeptidyl peptidase-IV (DPP-IV) 1, in which the N-terminal His-Ala is catalyzed cleavage by DPP-IV. HAEGT can be used in the research of diabetes, obesity.
Tekniske data
Renhed 99.26%
CAS 852155-81-8
Indhold og opbevaring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molekylvægt (g/mol) 513.5 g/mol
Produkttype Peptides