missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ HA 130
GREENER_CHOICE

Artikelnummer. 18035432
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18035432 50 mg
18014152 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18035432 Leverandør Tocris Bioscience™ Leverandørnr. 4196/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective autotaxin inhibitor

Potent and selective autotaxin inhibitor (IC50 = 28 nM). Binds and inhibits autotaxin reversibly. Displays no activity against the proteasome, recombinant NPP1 or alkaline phosphatase. Inhibits ATX-mediated cell migration in an A2058 melanoma cell assay.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229652-21-4
Molekylær formel C24H19BFNO5S

Specifications

Kemisk navn eller materiale HA 130
Mængde 50 mg
Formel vægt 463.29
Renhed >98%
Molekylvægt (g/mol) 463.286
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.