Få mere at vide
Tocris Bioscience™ HA 130
GREENER_CHOICE
Artikelnummer. 18014152
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18014152 10 mg
18035432 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18014152 Leverandør Tocris Bioscience™ Leverandørnr. 4196/10
Potent and selective autotaxin inhibitor
Potent and selective autotaxin inhibitor (IC50 = 28 nM). Binds and inhibits autotaxin reversibly. Displays no activity against the proteasome, recombinant NPP1 or alkaline phosphatase. Inhibits ATX-mediated cell migration in an A2058 melanoma cell assay.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1229652-21-4
Molekylær formel C24H19BFNO5S
Tekniske data
Kemisk navn eller materiale HA 130
Mængde 10 mg
Formel vægt 463.29
Renhed >98%
Molekylvægt (g/mol) 463.286