missing translation for 'onlineSavingsMsg'
Få mere at vide

H2N-Dpeg(4)-COOH Hydrochloride, TRC

Artikelnummer. 30395829
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30395829 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30395829 Leverandør Toronto Research Chemicals Leverandørnr. TRCH289595500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale H2N-Dpeg(4)-COOH Hydrochloride
CAS 2708292-39-9
Molekylær formel C11H23NO6.HCl
Mængde 500 mg
Synonym 15-Amino-4,7,10,13-tetraoxapentadecanoic acid Hydrochloride, 3-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]propionic acid Hydrochloride, 3-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]propanoic acid Hydrochloride
InChI formel InChI=1S/C11H23NO6.ClH/c12-2-4-16-6-8-18-10-9-17-7-5-15-3-1-11(13)14;/h1-10,12H2,(H,13,14);1H
SMIL O=C(CCOCCOCCOCCOCCN)O.Cl
IUPAC navn 3-[2-[2-[2-(2-aminoethoxy)ethoxy]ethoxy]ethoxy]propanoic acid;hydrochloride
Molekylvægt (g/mol) 301.76
Formel vægt 301.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.