Få mere at vide
H-Ser-Phe-OH, TRC
Artikelnummer. 30341006
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30341006 5 mg
1 options
Artikelnummer. 30341006 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7740935MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale H-Ser-Phe-OH
CAS 16875-28-8
Molekylær formel C12 H16 N2 O4
Mængde 5 mg
Synonym L-Seryl-L-phenylalanine, 100: PN: WO2020097235 SEQID: 330 Claimed Protein, 124: PN: US20130123467 SEQID: 149 Claimed Protein, 20: PN: US20090239809 SEQID: 20 Claimed Protein, 3: PN: WO2014063098 TABLE: 5 Claimed Sequence, 445: PN: WO2005081628 SEQID: 447 Claimed Protein, 7: PN: WO2011050471 SEQID: 7 Claimed Sequence, Serylphenylalanine
InChI formel InChI=1S/C12H16N2O4/c13-9(7-15)11(16)14-10(12(17)18)6-8-4-2-1-3-5-8/h1-5,9-10,15H,6-7,13H2,(H,14,16)(H,17,18)/t9-,10-/m0/s1
SMIL N[C@@H](CO)C(=O)N[C@@H](Cc1ccccc1)C(=O)O
IUPAC navn (2S)-2-[[(2S)-2-amino-3-hydroxypropanoyl]amino]-3-phenylpropanoic acid
Molekylvægt (g/mol) 252.266
Formel vægt 252.111