missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-Ser-Phe-OH, TRC
Specifications
Specifications
| Kemisk navn eller materiale | H-Ser-Phe-OH |
| CAS | 16875-28-8 |
| Molekylær formel | C12 H16 N2 O4 |
| Mængde | 10 mg |
| Synonym | L-Seryl-L-phenylalanine, 100: PN: WO2020097235 SEQID: 330 Claimed Protein, 124: PN: US20130123467 SEQID: 149 Claimed Protein, 20: PN: US20090239809 SEQID: 20 Claimed Protein, 3: PN: WO2014063098 TABLE: 5 Claimed Sequence, 445: PN: WO2005081628 SEQID: 447 Claimed Protein, 7: PN: WO2011050471 SEQID: 7 Claimed Sequence, Serylphenylalanine |
| InChI formel | InChI=1S/C12H16N2O4/c13-9(7-15)11(16)14-10(12(17)18)6-8-4-2-1-3-5-8/h1-5,9-10,15H,6-7,13H2,(H,14,16)(H,17,18)/t9-,10-/m0/s1 |
| SMIL | N[C@@H](CO)C(=O)N[C@@H](Cc1ccccc1)C(=O)O |
| IUPAC navn | (2S)-2-[[(2S)-2-amino-3-hydroxypropanoyl]amino]-3-phenylpropanoic acid |
| Molekylvægt (g/mol) | 252.266 |
| Formel vægt | 252.111 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?