Få mere at vide
H-Met-Asp-OH, TRC
Artikelnummer. 30476343
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476343 100 mg
30482449 10 mg
30480446 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476343 Leverandør Toronto Research Chemicals Leverandørnr. TRCM222188100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14595-65-4
Molekylær formel C9H16N2O5S
Molekylvægt (g/mol) 264.3
IUPAC navn (2S)-2-[[(2S)-2-amino-4-methylsulfanylbutanoyl]amino]butanedioic acid
SMIL CSCC[C@H](N)C(=O)N[C@@H](CC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale H-Met-Asp-OH
InChI formel InChI=1S/C9H16N2O5S/c1-17-3-2-5(10)8(14)11-6(9(15)16)4-7(12)13/h5-6H,2-4,10H2,1H3,(H,11,14)(H,12,13)(H,15,16)/t5-,6-/m0/s1
Mængde 100 mg
Formel vægt 264.078