Få mere at vide
H-Leu-val-oh, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | H-Leu-val-oh |
| CAS | 13588-95-9 |
| Molekylær formel | C11H22N2O3 |
| Mængde | 100 mg |
| Synonym | L-Leucyl-L-valine, 128: PN: WO2014102101 SEQID: 428 Claimed Protein, 128: PN: WO2014102104 SEQID: 227 Claimed Protein, 147: PN: WO2014102100 TABLE: 1 Claimed Protein, 168: PN: WO2014102103 TABLE: 1 Claimed Protein, 203: PN: EP2161028 PAGE: 11 Claimed Protein, 2095: PN: EP2071334 SEQID: 2206 Claimed Protein, 2095: PN: WO2009077864 SEQID: 2206 Claimed Protein, 20: PN: WO03052099 PAGE: 84 Claimed Protein, 30: PN: US20120167238 SEQID: 13 Claimed Sequence, 425: PN: WO03070760 PAGE: 99 Claimed Protein, 54: PN: WO2011044311 SEQID: 31 Unclaimed Protein, N-Leucylvaline |
| InChI formel | InChI=1S/C11H22N2O3/c1-6(2)5-8(12)10(14)13-9(7(3)4)11(15)16/h6-9H,5,12H2,1-4H3,(H,13,14)(H,15,16)/t8-,9-/m0/s1 |
| SMIL | CC(C)C[C@H](N)C(=O)N[C@@H](C(C)C)C(=O)O |
| Molekylvægt (g/mol) | 230.31 |
| Formel vægt | 230.163 |