Få mere at vide
(H-Hocys-ome)2 dihydrochloride, TRC
Artikelnummer. 30404530
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30404530 250 mg
1 options
Artikelnummer. 30404530 Leverandør Toronto Research Chemicals Leverandørnr. TRCH669603250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Halquinol, mixture of 5,7-Dichloro-8-quinolinol, 5-Chloro-8-quinolinol, 7-Chloro-8-quinolinol and 8-Hydroxyquinoline
CAS 8067-69-4
Molekylær formel C9 H7 N O . 2[C9 H6 Cl N O] . C9 H5 Cl2 N O
Mængde 250 mg
Synonym CHQ, Chlorhydroxyquinoline, Chlorquinol, Halquivet, Nutriquinol, Quixalud, SQ 16401Staquinol, Tarquinor
InChI formel InChI=1S/C9H5Cl2NO.2C9H6ClNO.C9H7NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8;10-7-3-4-8(12)9-6(7)2-1-5-11-9;10-7-4-3-6-2-1-5-11-8(6)9(7)12;11-8-5-1-3-7-4-2-6-10-9(7)8/h1-4,13H;2*1-5,12H;1-6,11H
SMIL Oc1cccc2cccnc12.Oc3ccc(Cl)c4cccnc34.Oc5c(Cl)ccc6cccnc56.Oc7c(Cl)cc(Cl)c8cccnc78
IUPAC navn 5-chloroquinolin-8-ol;7-chloroquinolin-8-ol;5,7-dichloroquinolin-8-ol;quinolin-8-ol