missing translation for 'onlineSavingsMsg'
Få mere at vide

H-D-Leu-L-Leu-OH, TRC

Artikelnummer. 30482315
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482315 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30482315 Leverandør Toronto Research Chemicals Leverandørnr. TRCL33080325MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale H-D-Leu-L-Leu-OH
CAS 38689-31-5
Molekylær formel C12H24N2O3
InChI formel InChI=1S/C12H24N2O3/c1-7(2)5-9(13)11(15)14-10(12(16)17)6-8(3)4/h7-10H,5-6,13H2,1-4H3,(H,14,15)(H,16,17)/t9-,10+/m1/s1
SMIL CC(C)C[C@@H](N)C(=O)N[C@@H](CC(C)C)C(=O)O
Molekylvægt (g/mol) 244.33
Mængde 25 mg
Formel vægt 244.1787
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.