Få mere at vide
GW311616 hydrochloride, MedChemExpress
Beskrivelse
GW-311616 is a potent, orally bioavailable, long duration and selective human neutrophil elastase (HNE) inhibitor with IC50 value of 22 nM and Ki value of 0.31 nM.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | GW311616 hydrochloride |
| CAS | 197890-44-1 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C19H32ClN3O4S |
| Mængde | 5 mg |
| Synonym | GW311616A |
| Opløselighedsinformation | DMSO : 100 mg/mL (230.42 mM; Need ultrasonic) ∣ H2O : 100 mg/mL (230.42 mM; Need ultrasonic) |
| SMIL | O=C1[C@@H](C(C)C)[C@@](N(C(/C=C/CN2CCCCC2)=O)CC3)([H])[C@]3([H])N1S(=O)(C)=O.[H]Cl |
| Molekylvægt (g/mol) | 433.99 |
| Vis mere |