missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
GW 766994, MedChemExpress
Beskrivelse
GW 766994 (GW 994) is an orally active and specific chemokine receptor-3 (CCR3) antagonist. GW 766994 has the potential for asthma and eosinophilic bronchitis research.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | GW 766994 |
| Fysisk form | Solid |
| CAS | 408303-43-5 |
| Farve | Light Yellow |
| Mængde | 10 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C21H24Cl2N4O3 |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | GW 994 |
| Opløselighedsinformation | DMSO : 56 mg/mL (124.07 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?