missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ GW 1929 hydrochloride
GREENER_CHOICE

Artikelnummer. 18066022
Change view
Click to view available options
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18066022 10 mg
18014682 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18066022 Supplier Tocris Bioscience™ Supplier No. 1664/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective PPARγ agonist. Orally active

Highly selective orally active peroxisome proliferator-activated receptor (PPAR)γ agonist (pEC50 values are 8.05, < 4 and < 4 for human PPARγ, PPARα and PPARδ receptors respectively). Decreases glucose, fatty acid and triglyceride levels following oral administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217466-21-1
Molekylær formel C30H30ClN3O4

Specifications

Kemisk navn eller materiale GW 1929 hydrochloride
Mængde 10 mg
Formel vægt 532.03
Renhed >98%
Molekylvægt (g/mol) 532.037
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.