Få mere at vide
Gusacitinib, MedChemExpress
Artikelnummer. 16420157
Encompass_Preferred
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
16420157 50 mg
1 options
Artikelnummer. 16420157 Leverandør Medchem Express Leverandørnr. HY103018
MedChemExpress Gusacitinib (ASN-002) is an orally active and potent dual inhibitor of spleen tyrosine kinase (SYK) and janus kinase (JAK) with IC50 values of 5-46 nM. Gusacitinib has anti-cancer activity in both solid and hematological tumor types.
Gusacitinib (ASN-002) is an orally active and potent dual inhibitor of spleen tyrosine kinase (SYK) and janus kinase (JAK) with IC50 values of 5-46 nM. Gusacitinib has anti-cancer activity in both solid and hematological tumor types.
Tekniske data
Kemisk navn eller materiale Gusacitinib
CAS 1425381-60-7
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C24H28N8O2
Mængde 50 mg
Synonym ASN-002
Opløselighedsinformation DMSO : 100 mg/mL (217.14 mM; Need ultrasonic)
SMIL N#CCC1CCN(C2=NC(NC3=CC=C(N4CCC(O)CC4)C=C3)=C5C(C=NNC5=O)=N2)CC1
Molekylvægt (g/mol) 460.53
Formel vægt 460.53
Procent renhed 98.59%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease