Få mere at vide
Guanoxan Sulfate, LoGiCal
Artikelnummer. 30460987
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460987 10 mg
1 options
Artikelnummer. 30460987 Leverandør LoGiCal Leverandørnr. LGCFOR1880.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Guanoxan Sulfate
CAS 4-5-5714
Analyt- eller komponentnavne Guanoxan Sulfate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C10 H13 N3 O2 . H2 O4 S
Certificeringer/compliance ISO 9001
Synonym Guanidine, N-[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-, sulfate (2:1), Guanidine, (1,4-benzodioxan-2-ylmethyl)-, sulfate (2:1) (8CI), Guanidine, [(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-, sulfate (2:1) (9CI), Envacar, Guanoxan sulfate, Guanutil, NSC 108162, P 1003, P 3/01003
InChI formel InChI=1S/2C10H13N3O2.H2O4S/c2*11-10(12)13-5-7-6-14-8-3-1-2-4-9(8)15-7;1-5(2,3)4/h2*1-4,7H,5-6H2,(H4,11,12,13);(H2,1,2,3,4)
Løsningstype Neat
SMIL NC(=N)NCC1COc2ccccc2O1.NC(=N)NCC3COc4ccccc4O3.OS(=O)(=O)O
IUPAC navn 1-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)guanidine;sulfuric acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 512.54
Mængde 10 mg
Formel vægt 512.1689 g/mol