Få mere at vide
MedChemExpress Guanosine 5'-triphosphate trisodium salt hydrate
Tekniske data
Tekniske data
| Produkttype | Reference compound |
| Indhold og opbevaring | -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Forsendelsestilstand | Room temperature |
| Mængde | 200 mg |
| Opløselighedsinformation | H2O : ≥ 150 mg/mL |
| CAS | 207300-85-4 |
| Synonym | 5'-GTP trisodium salt hydrate |