missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanosine-5'-Triphosphate Disodium Salt, Thermo Scientific™
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Plastic bottle
Specifications
Physical Form | Crystalline Powder |
Quantity | 100 mg |
Beilstein | 4113439 |
Synonym | guanosine 5 '-triphosphate, guanosine-5 '-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5 '-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate |
Formula Weight | 567.14 |
Packaging | Plastic bottle |
Chemical Name or Material | Guanosine-5 '-Triphosphate Disodium Salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'