Få mere at vide
Tocris Bioscience™ Guangxitoxin 1E
Potent Kv2.1 and Kv2.2 channel blocker
Brand: Tocris Bioscience™ 5676/100U
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Guangxitoxin 1E is a Kv2.1 and Kv2.2 channel blocker (IC50 values are 1-3 nM). Enhances glucose-stimulated insulin secretion from human islets in vitro, but not from islet cells lacking the Kv2.1 channel. Has no significant effect on plasma insulin, glucagon or blood glucose levels in mice, but increases plasma somatostatin levels.Tekniske data
| 1233152-82-3 | |
| Voltage-gated Potassium Channel Blockers | |
| 100 μg |
| Store at -20°C | |
| Observed M.W.: 3948.61 | |
| C178H248N44O45S7 |