Få mere at vide
Guaiacol Rutinoside, TRC
Artikelnummer. 30008021
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008021 1 mg
30004187 10 mg
30005449 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008021 Leverandør Toronto Research Chemicals Leverandørnr. TRCG2455801MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1416253-69-4
Molekylær formel C19 H28 O11
Molekylvægt (g/mol) 432.42
Synonym 2-Methoxyphenyl 6-O-(6-deoxy-a-L-mannopyranosyl)-β -D-glucopyranoside
IUPAC navn (2S,3R,4R,5R,6R)-2-methyl-6-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-methoxyphenoxy)tetrahydropyran-2-yl]methoxy]tetrahydropyran-3,4,5-triol
SMIL COc1ccccc1O[C@@H]2O[C@H](CO[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O
Tekniske data
Kemisk navn eller materiale Guaiacol Rutinoside
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C19H28O11/c1-8-12(20)14(22)16(24)18(28-8)27-7-11-13(21)15(23)17(25)19(30-11)29-10-6-4-3-5-9(10)26-2/h3-6,8,11-25H,7H2,1-2H3/t8-,11+,12-,13+,14+,15-,16+,17+,18+,19+/m0/s1
Formel vægt 432.1632
Procent renhed >95%