Learn More
Cytiva GST Gene Fusion System pGEX Vectors
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
5008.00 DKK
Specifications
Quantity | 25 μg |
---|---|
For Use With (Application) | Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST |
Type | pGEX Vector |
Product Code | Brand | Kit Contents | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Kit Contents | Price | Quantity & Availability | |||||
10534435
|
Cytiva
28-9545-49 |
4T-1 |
5008.00 DKK
25µL |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10209814
|
Cytiva
28-9546-50 |
6P-2 |
5008.00 DKK
25µL |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10516355
|
Cytiva
28-9546-51 |
6P-3 |
5008.00 DKK
25µL |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10350355
|
Cytiva
28-9546-48 |
6P-1 |
5008.00 DKK
25µL |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10437484
|
Cytiva
28-9546-53 |
2T |
5008.00 DKK
25µg |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10340735
|
Cytiva
28-9545-52 |
4T-3 |
5008.00 DKK
25µL |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
Description
- Offers a tac promoter for chemically inducible, high-level expression of GST-tagged recombinant proteins
- Offers an internal laq1q gene for use in any E. coli host
- With PreScission™ Protease, Thrombin, or Factor Xa recognition sites for cleaving the target protein from the fusion product
- The vectors provide all three translational reading frames beginning with the EcoRI restriction site
- Can use a wide variety of E. coli host strains cloning and expression
- pGEX-1lT EcoRI/BAP is an EcoR1 pre cut vector, phosphatase treated to prevent self ligation, ready for gene insertion
- pGEX-2TK will introduce a specific kinase site allowing the fusion protein to be labelled with [γ-32P]ATP using protein kinase
Specifications
25 μg | |
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST | |
pGEX Vector |