missing translation for 'onlineSavingsMsg'
Learn More
GSK805, MedChemExpress Product Code.: 16475338

GSK805, MedChemExpress

Product Code. p-200097661
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475338

Brand: Medchem Express HY12776

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK805 is an orally active and CNS penetrant RORÎłt inhibitor. GSK805 inhibits RORÎł and Th17 cells differentiation with pIC50 values of 8.4 and >8.2. GSK805 inhibits the function of Th17 cells. GSK805 can be used for the research of immunity.

GSK805 is an orally active and CNS penetrant RORÎłt inhibitor. GSK805 inhibits RORÎł and Th17 cells differentiation with pIC50 values of 8.4 and >8.2. GSK805 inhibits the function of Th17 cells. GSK805 can be used for the research of immunity.

Specifications

Chemical Name or Material GSK805
CAS 1426802-50-7
Color White
Purity Grade Notes Research
Molecular Formula C23H18Cl2F3NO4S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (187.84 mM)
SMILES O=C(NC1=CC(Cl)=C(C2=CC=CC=C2OC(F)(F)F)C(Cl)=C1)CC3=CC=C(S(=O)(CC)=O)C=C3
Molecular Weight (g/mol) 532.36
Formula Weight 532.36
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
GSK805, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.