missing translation for 'onlineSavingsMsg'
Learn More
GSK583, MedChemExpress Product Code.: 16460006

GSK583, MedChemExpress

Product Code. p-200097243
2 mg, 2mg
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460006

Brand: Medchem Express HY100339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK583 is a highly potent, orally active and selective inhibitor of RIP2 Kinase, with IC50 of 5 nM. GSK583 inhibits both TNF-α and IL-6 production with an IC50 value of 200 nM.

GSK583 is a highly potent, orally active and selective inhibitor of RIP2 Kinase, with IC50 of 5 nM. GSK583 inhibits both TNF-α and IL-6 production with an IC50 value of 200 nM.

Specifications

Chemical Name or Material GSK583
CAS 1346547-00-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H19FN4O2S
Quantity 2 mg
Solubility Information DMSO : ≥ 100 mg/mL (250.97 mM)
SMILES FC1=CC2=C(NN=C2NC3=CC=NC4=CC=C(S(=O)(C(C)(C)C)=O)C=C34)C=C1
Molecular Weight (g/mol) 398.45
Formula Weight 398.45
Percent Purity 98.74%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
GSK583, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.