missing translation for 'onlineSavingsMsg'
Learn More

GSK3368715 dihydrochloride, MedChemExpress

Product Code. 16491309
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16491309 1 mg -
16458137 5 mg -
16441607 50 mg -
16415428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491309 Supplier Medchem Express Supplier No. HY128717A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK3368715 dihydrochloride (EPZ019997 dihydrochloride) is an orally active, reversible, and S-adenosyl-L-methionine (SAM) uncompetitive type I protein arginine methyltransferases (PRMTs) inhibitor (IC50=3.1 nM (PRMT1), 48 nM (PRMT3), 1148 nM (PRMT4), 5.7 nM (PRMT6), 1.7 nM (PRMT8)).

GSK3368715 dihydrochloride (EPZ019997 dihydrochloride) produces a shift in arginine methylation states, alters exon usage, and has strong anti-cancer activity.

Specifications

Kemisk navn eller materiale GSK3368715 dihydrochloride
Fysisk form Powder
CAS 1628925-77-8
Farve White
Mængde 1 mg
Renhedsgrad noter Research
Molekylær formel C20H40Cl2N4O2
Til brug med (applikation) Cancer-programmed cell death
Synonym EPZ019997 dihydrochloride
Opløselighedsinformation DMSO : 230 mg/mL (523.37 mM; Need ultrasonic) ∣H2O : 140 mg/mL (318.57 mM; Need ultrasonic)
SMIL CNCCN(CC1=CNN=C1C2CCC(COCC)(COCC)CC2)C.Cl.Cl
Molekylvægt (g/mol) 439.46
Formel vægt 439.46
Procent renhed 99.66%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.