Få mere at vide
GSK-LSD1 dihydrochloride, MedChemExpress
Artikelnummer. 16492606
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16492606 25 mg
16489636 50 mg
16463078 10 mg
16405817 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16492606 Leverandør Medchem Express Leverandørnr. HY100546A
MedChemExpress GSK-LSD1 dihydrochloride is a potent, selective and irreversible lysine specific demethylase 1 (LSD1) inhibitor with an IC50 of 16 nM.
GSK-LSD1 dihydrochloride is a potent, selective and irreversible lysine specific demethylase 1 (LSD1) inhibitor with an IC50 of 16 nM.
Tekniske data
Kemisk navn eller materiale GSK-LSD1 dihydrochloride
CAS 2102933-95-7
Farve White
Renhedsgrad noter Research
Molekylær formel C14H22Cl2N2
Mængde 25 mg
Opløselighedsinformation DMSO : ≥ 62.5 mg/mL (216.08 mM)
SMIL C1(N[C@H]2[C@H](C3=CC=CC=C3)C2)CCNCC1.Cl.Cl
Molekylvægt (g/mol) 289.24
Formel vægt 289.24
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death