Få mere at vide
Tocris Bioscience™ GSK LSD 1 Dihydrochloride
Potent and selective LSD1 inhibitor
Brand: Tocris Bioscience™ 5361/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GSK LSD 1 dihydrochloride is a potent and selective lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 16 nM). Exhibits >1000-fold selectivity for LSD1 over closely related enzymes including LSD2, MAO-1 and MAO-B. Inhibits cancer cell line growth in vitro.Tekniske data
| GSK LSD 1 dihydrochloride | |
| rel-N-[(1S,2R)-2-Phenylcyclopropyl]-4-piperidinamine dihydrochloride | |
| Soluble to 100mM in water and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 2102933-95-7 | |
| 10 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use