Få mere at vide
Tocris Bioscience™ GSK J1
Artikelnummer. 18034352
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18034352 10 mg
18055332 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18034352 Leverandør Tocris Bioscience™ Leverandørnr. 4593/10
Potent JMJD3/UTX inhibitor
Potent inhibitor of the H3K27 histone demethylases JMJD3 (KDM6B) and UTX (KDM6A) (IC50 values are 28 and 53 nM respectively). Also inhibits KDM5B, KDM5C and KDM5A (IC50 values are 170, 550 and 6,800 nM respectively). Exhibits no activity against a panel of other histone demethylases (IC50 >20 μM), and displays no significant inhibitory activity against 100 protein kinases at a concentration of 30 μM.
Ethyl ester derivative and Negative Control also available.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1373422-53-7
Molekylær formel C22H23N5O2
Tekniske data
Kemisk navn eller materiale GSK J1
Mængde 10 mg
Formel vægt 389.45
Renhed >99%
Molekylvægt (g/mol) 389.459