missing translation for 'onlineSavingsMsg'
Få mere at vide

GSK-872, MedChemExpress

Artikelnummer. 16490017
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16490017 50 mg -
16472796 25 mg -
16463628 10 mg -
16436357 5 mg -
16414716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16490017 Leverandør Medchem Express Leverandørnr. HY101872

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1.3 nM. GSK-872 decreases the RIPK3-mediated necroptosis and subsequent cytoplasmic translocation and expression of HMGB1, as well as ameliorates brain edema and neurological deficits in early brain injury.

GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1.3 nM. GSK-872 decreases the RIPK3-mediated necroptosis and subsequent cytoplasmic translocation and expression of HMGB1, as well as ameliorates brain edema and neurological deficits in early brain injury.

Tekniske data

Kemisk navn eller materiale GSK-872
CAS 1346546-69-7
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C19H17N3O2S2
Mængde 50 mg
Opløselighedsinformation DMSO : 100 mg/mL (260.76 mM; Need ultrasonic)
SMIL O=S(C1=CC=C2N=CC=C(C2=C1)NC3=CC=C4SC=NC4=C3)(C(C)C)=O
Molekylvægt (g/mol) 383.49
Formel vægt 383.49
Procent renhed 98.0%
Grad Research
Fysisk form Powder
Til brug med (applikation) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.