Få mere at vide
Tocris Bioscience™ GSK 343
Potent and selective SAM-competitive EZH2 inhibitor
Brand: Tocris Bioscience™ 6128/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
GSK 343 is a potent and selective SAM-competitive EZH2 inhibitor (IC50 = 4 nM), that exhibits >60 fold selectivity for EZH2 over EZH1 and a range of other methyltransferases. GSK 343 decreases H3K27me3 levels in breast cancer cells and inhibits proliferation of prostate cancer cell lines in vitro.Tekniske data
| N-[(1,2-Dihydro-6-methyl-2-oxo-4-propyl-3-pyridinyl)methyl]-1-(1-methylethyl)-6-[2-(4-methyl-1-piperazinyl)-4-pyridinyl]-1H-indazole-4-carboxamide | |
| 1346704-33-3 | |
| >98% | |
| 10 mg |