Få mere at vide
Tocris Bioscience™ GSK 343
GREENER_CHOICE
Potent and selective SAM-competitive EZH2 inhibitor
Brand:  Tocris Bioscience™ 6128/10
Encompass
Produktkode 15726369
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
GSK 343 is a potent and selective SAM-competitive EZH2 inhibitor (IC50 = 4 nM), that exhibits >60 fold selectivity for EZH2 over EZH1 and a range of other methyltransferases. GSK 343 decreases H3K27me3 levels in breast cancer cells and inhibits proliferation of prostate cancer cell lines in vitro.
Tekniske data
Tekniske data
N-[(1,2-Dihydro-6-methyl-2-oxo-4-propyl-3-pyridinyl)methyl]-1-(1-methylethyl)-6-[2-(4-methyl-1-piperazinyl)-4-pyridinyl]-1H-indazole-4-carboxamide
1346704-33-3
>98%
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner